Order Entry
Italy
ContactUsLinkComponent
Anti-Dysferlin Rabbit Monoclonal Antibody [clone: JAI-1-49-3]
Anti-Dysferlin Rabbit Monoclonal Antibody [clone: JAI-1-49-3]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-Dysferlin Rabbit Monoclonal Antibody [clone: JAI-1-49-3]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCAAB124684-100
  • ABCAAB124684-1ML
  • Antigen symbol:
  • DYSF
  • Antigen name:
  • Dysferlin
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • JAI-1-49-3
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • Western blot:
  • Yes
  • ImmunoChemistry:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

Specifications

Frequently Bought Together