Order Entry
Italy
ContactUsLinkComponent
Anti-Soluble TNF Receptor II Rabbit Polyclonal Antibody
Anti-Soluble TNF Receptor II Rabbit Polyclonal Antibody
  ABCAAB109853-50
 :  Abcam
undefined
Anti-Soluble TNF Receptor II Rabbit Polyclonal Antibody
  ABCAAB109853-50
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    Soluble TNF Receptor II
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    1202
  • Neutralisation:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pack type:
    Vial
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: Soluble TNF Receptor II
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human