Order Entry
Italy
ContactUsLinkComponent
Anti-AMPK alpha 2 (phospho S491) Rabbit Monoclonal Antibody [clone: EPR3052]
Anti-AMPK alpha 2 (phospho S491) Rabbit Monoclonal Antibody [clone: EPR3052]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-AMPK alpha 2 (phospho S491) Rabbit Monoclonal Antibody [clone: EPR3052]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCAAB109402-100
  • ABCAAB109402-1ML
  • Antigen symbol:
  • PRKAA2
  • Antigen name:
  • Protein kinase, AMP-activated, alpha 2 catalytic subunit
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • EPR3052
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • Western blot:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

Specifications

Frequently Bought Together