Order Entry
Italy
ContactUsLinkComponent
Anti-ABCG2 Rabbit Monoclonal Antibody [clone: EPR2099]
Anti-ABCG2 Rabbit Monoclonal Antibody [clone: EPR2099]
  ABCAAB108312-100
 :  Abcam
Anti-ABCG2 Rabbit Monoclonal Antibody [clone: EPR2099]
  ABCAAB108312-100
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    ATP Binding Cassette, sub-family G, member 2
  • Antigen symbol:
    ABCG2
  • Clonality:
    Monoclonal
  • Clone:
    EPR2099
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ABCG2 (ATP-binding cassette, sub-family G (WHITE), member 2)
Clonality: monoclonal
Clone: EPR2099
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: