Order Entry
Italy
ContactUsLinkComponent
 
Anti-EVI2B Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-216]
  ABCAAB101040-50
 :  Abcam
Anti-EVI2B Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-216]
  ABCAAB101040-50
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    ecotropic viral integration site 2B
  • Antigen symbol:
    EVI2B
  • Clonality:
    Monoclonal
  • Clone:
    MEM-216
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 Tests
  • Pk:
    50 Tests

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: EVI2B (ecotropic viral integration site 2B)
Clonality: monoclonal
Clone: MEM-216
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human