Order Entry
Italy
ContactUsLinkComponent
Anti-CTGF Mouse monoclonal antibody unconjugated [clone: 17-73-F1]
Anti-CTGF Mouse monoclonal antibody unconjugated [clone: 17-73-F1]
Catalog # ABCAAB94939-100
Supplier:  Abcam
Anti-CTGF Mouse monoclonal antibody unconjugated [clone: 17-73-F1]
Catalog # ABCAAB94939-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    connective tissue growth factor
  • Antigen symbol:
    CTGF
  • Clonality:
    Monoclonal
  • Clone:
    17-73-F1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CTGF (connective tissue growth factor)
Clonality: monoclonal
Clone: 17-73-F1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human