Anti-SDF4 Rabbit Polyclonal Antibody
Catalog #
Product Details & Documents
Type: Primary
Antigen: SDF4 (stromal cell derived factor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Type: Primary
Antigen: SDF4 (stromal cell derived factor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human