Order Entry
Italy
ContactUsLinkComponent
 
Anti-Phospholipase D1 Rabbit Monoclonal Antibody [clone: EP1506Y]
  ABCAAB68150-100
 :  Abcam
undefined
Anti-Phospholipase D1 Rabbit Monoclonal Antibody [clone: EP1506Y]
  ABCAAB68150-100
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    Phospholipase D1, phosphatidylcholine-specific
  • Antigen symbol:
    PLD1
  • Clonality:
    Monoclonal
  • Clone:
    EP1506Y
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Rabbit monoclonal [EP1506Y] to Phospholipase D1.

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PLD1 (phospholipase D1, phosphatidylcholine-specific)
Clonality: monoclonal
Clone: EP1506Y
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: