Order Entry
Italy
ContactUsLinkComponent
Anti-SCN4A Rabbit Polyclonal Antibody
Anti-SCN4A Rabbit Polyclonal Antibody
Catalog # ABCAAB65165-100
Supplier:  Abcam
undefined
Anti-SCN4A Rabbit Polyclonal Antibody
Catalog # ABCAAB65165-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    sodium channel, voltage-gated, type IV, alpha subunit
  • Antigen symbol:
    SCN4A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal to SCN4A.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SCN4A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human