Order Entry
Italy
ContactUsLinkComponent
 
Anti-Interferon alpha/beta receptor 1 Rabbit Polyclonal Antibody
  ABCAAB62693-100
 :  Abcam
undefined
Anti-Interferon alpha/beta receptor 1 Rabbit Polyclonal Antibody
  ABCAAB62693-100
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    Interferon (alpha, beta and omega) receptor 1
  • Antigen symbol:
    IFNAR1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

 

 

Rabbit polyclonal to Interferon alpha/beta receptor 1.

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Interferon alpha/beta receptor 1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human