Order Entry
Italy
ContactUsLinkComponent
 
Anti-Integrin beta 3 Armenian hamster Monoclonal Antibody [clone: HM beta 3.1] (PE (Phycoerythrin))
  ABCAAB54190-50
 :  Abcam
undefined
Anti-Integrin beta 3 Armenian hamster Monoclonal Antibody [clone: HM beta 3.1] (PE (Phycoerythrin))
  ABCAAB54190-50
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61)
  • Antigen symbol:
    ITGB3
  • Clonality:
    Monoclonal
  • Clone:
    HM beta 3.1
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Armenian hamster
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 tests
  • Pack type:
    Vial
  • Pk:
    50 Tests

 

 

PE Armenian hamster monoclonal [HM beta 3.1] to Integrin beta 3.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Integrin beta 3
Clonality: Monoclonal
Clone: HM beta 3.1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian hamster
Isotype: IgG
Reactivity: Mouse;Rat