Order Entry
Italy
ContactUsLinkComponent
Anti-Cytokeratin 18 Mouse Monoclonal Antibody [clone: C-04] (FITC (Fluorescein Isothiocyanate))
Anti-Cytokeratin 18 Mouse Monoclonal Antibody [clone: C-04] (FITC (Fluorescein Isothiocyanate))
Catalog # ABCAAB52459-100
Supplier:  Abcam
undefined
Anti-Cytokeratin 18 Mouse Monoclonal Antibody [clone: C-04] (FITC (Fluorescein Isothiocyanate))
Catalog # ABCAAB52459-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    keratin 18
  • Antigen symbol:
    KRT18
  • Clonality:
    Monoclonal
  • Clone:
    C-04
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

Specifications

About this item

FITC Mouse monoclonal [C-04] to Cytokeratin 18.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cytokeratin 18
Clonality: Monoclonal
Clone: C-04
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human