Order Entry
Italy
ContactUsLinkComponent
Anti-CXCL14 Rabbit Polyclonal Antibody (Biotin)
Anti-CXCL14 Rabbit Polyclonal Antibody (Biotin)
Catalog # ABCAAB46008-50
Supplier:  Abcam
Anti-CXCL14 Rabbit Polyclonal Antibody (Biotin)
Catalog # ABCAAB46008-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    chemokine (C-X-C motif) ligand 14
  • Antigen symbol:
    CXCL14
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CXCL14 (chemokine (C-X-C motif) ligand 14)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human