Order Entry
Italy
ContactUsLinkComponent
Anti-CD18 Mouse Monoclonal Antibody [clone: MEM-48] (APC (Allophycocyanin))
Anti-CD18 Mouse Monoclonal Antibody [clone: MEM-48] (APC (Allophycocyanin))
Catalog # ABCAAB40914-50
Supplier:  Abcam
undefined
Anti-CD18 Mouse Monoclonal Antibody [clone: MEM-48] (APC (Allophycocyanin))
Catalog # ABCAAB40914-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    integrin, beta 2 (complement component 3 receptor 3 and 4 subunit)
  • Antigen symbol:
    ITGB2
  • Clonality:
    Monoclonal
  • Clone:
    MEM-48
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Seq:
    0780
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 Tests
  • Pack type:
    Vial
  • Pk:
    50 Tests

Specifications

About this item

APC Mouse monoclonal [MEM-48] to CD18, prediluted.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD18
Clonality: Monoclonal
Clone: MEM-48
Conjugation: APC (Allophycocyanin)
Epitope: This antibody recognizes an epitope including amino acids 534 - 546 in the cysteine - rich repeat 3 of the CD18 antigen
Host: Mouse
Isotype: IgG1
Reactivity: Human