Order Entry
Italy
ContactUsLinkComponent
Anti-SIRP alpha Mouse Monoclonal Antibody [clone: OX-41] (FITC (Fluorescein Isothiocyanate))
Anti-SIRP alpha Mouse Monoclonal Antibody [clone: OX-41] (FITC (Fluorescein Isothiocyanate))
Catalog # ABCAAB34076-50
Supplier:  Abcam
undefined
Anti-SIRP alpha Mouse Monoclonal Antibody [clone: OX-41] (FITC (Fluorescein Isothiocyanate))
Catalog # ABCAAB34076-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    SIRP alpha
  • Clonality:
    Monoclonal
  • Clone:
    OX-41
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Rat
  • Seq:
    1202
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pack type:
    Vial
  • Pk:
    50 µG

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SIRP alpha
Clonality: monoclonal
Clone: OX-41
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat