Anti-STAT1 Rabbit Polyclonal Antibody
Product Details & Documents
Rabbit Polyclonal STAT1 antibody. Suitable for WB, IHC-P and reacts with Human samples. Cited in 30 publications. Immunogen corresponding to Synthetic Peptide within Human STAT1 aa 650 to C-terminus.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: STAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Seq:1286
- Size:100 µg
- Pack type:Vial
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Immunogen Affinity purification
- Antibody type:Primary
- Form:Liquid
- Isotype:IgG
- Storage temperature:Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Host:Rabbit
- Concentration:1 mg/ml
- Molecular weight:87335 Da
- Shipping temperature:4 °C
- Antigen name:Signal transducer and activator of transcription 1-alpha/beta
- Cat. No.:ABCAAB31369-100
- Reactivity:Human
- Antigen synonyms:Transcription factor ISGF-3 components p91/p84
- Immunogen:Synthetic Peptide within Human STAT1 aa 650 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Tested applications:IHC-P
- Antigen symbol:STAT1
- Storage buffer:PBS, glycerol, sodium chloride
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



