Order Entry
Italy
ContactUsLinkComponent
Anti-CD8 alpha Rat Monoclonal Antibody [clone: 53-6.7] (FITC (Fluorescein Isothiocyanate))
Anti-CD8 alpha Rat Monoclonal Antibody [clone: 53-6.7] (FITC (Fluorescein Isothiocyanate))
Catalog # ABCAAB25676-50
Supplier:  Abcam
undefined
Anti-CD8 alpha Rat Monoclonal Antibody [clone: 53-6.7] (FITC (Fluorescein Isothiocyanate))
Catalog # ABCAAB25676-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD8a molecule
  • Antigen symbol:
    CD8A
  • Clonality:
    Monoclonal
  • Clone:
    53-6.7
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Mouse
  • Seq:
    1202
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pack type:
    Vial
  • Pk:
    50 µG

Specifications

About this item

FITC Rat monoclonal [53-6.7] to CD8 alpha.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD8 alpha
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse