Order Entry
Italy
ContactUsLinkComponent
Anti-MHC Class 1 H2 Db/H2-D1 Mouse Monoclonal Antibody [clone: 27-11-13] PE (Phycoerythrin)
Anti-MHC Class 1 H2 Db/H2-D1 Mouse Monoclonal Antibody [clone: 27-11-13] PE (Phycoerythrin)
Supplier:  Abcam
Certificates
undefined
Anti-MHC Class 1 H2 Db/H2-D1 Mouse Monoclonal Antibody [clone: 27-11-13] PE (Phycoerythrin)
Supplier:  Abcam

Specifications

  • Catalog No:
  • ABCAAB25547-100
  • ABCAAB25547-25
  • Antigen name:
  • MHC Class 1 H2 Db
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • Clone:
  • 27-11-13
  • Reactivity:
  • Mouse
  • Host:
  • Mouse
  • Isotype:
  • IgG2a
  • ImmunoChemistry:
  • Yes
  • Flow cytometry:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

Specifications

Frequently Bought Together