Order Entry
Italy
ContactUsLinkComponent
Anti-Scn1a Rabbit Polyclonal Antibody
Anti-Scn1a Rabbit Polyclonal Antibody
Catalog # ABCAAB24820-500
Supplier:  Abcam
undefined
Anti-Scn1a Rabbit Polyclonal Antibody
Catalog # ABCAAB24820-500
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    sodium channel, voltage-gated, type I, alpha subunit
  • Antigen symbol:
    SCN1A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Seq:
    1418
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    500 µl
  • Pack type:
    Vial
  • Pk:
    500 µl

Specifications

About this item

Rabbit polyclonal to Scn1a.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Scn1a
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat;Human