Anti-HLA Class I Mouse Monoclonal Antibody [clone: W6/32]
Supplier: Abcam
New Product
Certificates
About this item
Mouse monoclonal [W6/32] to HLA Class I - BSA and Azide free.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: HLA Class I
Clonality: Monoclonal
Clone: W6/32
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Specifications
- Catalog No:
- ABCAAB23755-1MG
- ABCAAB23755-100
- Tipo di anticorpi:
- HLA Class 1
- Tipo anticorpo:
- Primary
- Clonalità:
- Monoclonale
- Coniugazione:
- Non coniugato
- Clone:
- W6/32
- Ospite:
- Mouse
- Isotipo:
- IgG2a
- Western blot:
- Yes
- ELISA:
- Yes
- ImmunoChimica:
- Yes
- ImmunoPrecipitazione:
- Yes
- ImmunoFluorescenza:
- Yes
- Citometria a flusso:
- Yes
- Temperatura di conservazione:
- Cool
- Temperatura di spedizione:
- Ice
Specifications
Frequently Bought Together