- Antibody type:Primary
- Antigen name:anaplastic lymphoma receptor tyrosine kinase
- Antigen symbol:ALK
- Clonality:Monoclonal
- Clone:SP8
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:500 µl
- Pk:500 µl
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ALK (anaplastic lymphoma receptor tyrosine kinase)
Clonality: monoclonal
Clone: SP8
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: