Order Entry
Italy
ContactUsLinkComponent
Anti-EBNA 3A Sheep Polyclonal Antibody
Anti-EBNA 3A Sheep Polyclonal Antibody
Catalog # ABCAAB16126-250
Supplier:  Abcam
Anti-EBNA 3A Sheep Polyclonal Antibody
Catalog # ABCAAB16126-250
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    EBNA 3A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Sheep
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    250 µg
  • Pk:
    250 µG

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: EBNA 3A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: