Order Entry
Italy
ContactUsLinkComponent
Anti-CD146 Mouse Monoclonal Antibody [clone: OJ79c MUC 18]
Anti-CD146 Mouse Monoclonal Antibody [clone: OJ79c MUC 18]
Catalog # ABCAAB10816-50
Supplier:  Abcam
undefined
Anti-CD146 Mouse Monoclonal Antibody [clone: OJ79c MUC 18]
Catalog # ABCAAB10816-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Melanoma Cell Adhesion Molecule
  • Antigen symbol:
    MCAM
  • Clonality:
    Monoclonal
  • Clone:
    OJ79c MUC 18
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Seq:
    1202
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pack type:
    Vial
  • Pk:
    50 µG

Specifications

About this item

Mouse monoclonal [OJ79c MUC 18] to CD146.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD146
Clonality: Monoclonal
Clone: OJ79c MUC 18
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human