Order Entry
Italy
ContactUsLinkComponent
Anti-APC Mouse Monoclonal Antibody [clone: ALi 12-28]
Anti-APC Mouse Monoclonal Antibody [clone: ALi 12-28]
Catalog # ABCAAB58-100
Supplier:  Abcam
undefined
Anti-APC Mouse Monoclonal Antibody [clone: ALi 12-28]
Catalog # ABCAAB58-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    adenomatous polyposis coli
  • Antigen symbol:
    APC
  • Clonality:
    Monoclonal
  • Clone:
    ALi 12-28
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [ALi 12-28] to APC.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: APC (Allophycocyanin)
Clonality: Monoclonal
Clone: ALi 12-28
Conjugation:
Epitope: ALI-12-28 was epitope mapped by differential in vitro expression of the N-terminal region of the APC (Allophycocyanin) gene by using the protein truncation test and was found to bind to APC (Allophycocyanin) in the region between nucleotides 135 and 422 (exons 2-3) (Efstathiou et al.).
Host: Mouse
Isotype: IgG1
Reactivity: Human