Order Entry
Italy
ContactUsLinkComponent
Anti-Ras-related protein Rab-4 (RAB4) Rabbit Polyclonal Antibody
Anti-Ras-related protein Rab-4 (RAB4) Rabbit Polyclonal Antibody
Catalog # BIRBORB96512-50
Supplier:  Biorbyt
undefined
Anti-Ras-related protein Rab-4 (RAB4) Rabbit Polyclonal Antibody
Catalog # BIRBORB96512-50
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ras-related Protein Rab-4
  • Antigen symbol:
    RAB4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Western blot:
    Yes
  • Format:
    antiserum
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    C-terminal peptide from human Rab4
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Immunochemistry: 1:200; Western Blot: 1:1000-1:2000

Type: Primary
Antigen: Ras-related protein Rab-4 (RAB4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: