Anti-PRKAB1 Rabbit Polyclonal Antibody
BIRBORB96112-50
: Biorbyt
- Antibody type:Primary
- Antigen name:Protein Kinase AMP-Activated Non-Catalytic Subunit Beta 1
- Antigen symbol:PRKAB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Western blot:Yes
- Storage buffer:PBS, 0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
ELISA: 1:50-1:100; Immunochemistry: 10 ?g/ml; Western Blot: 1:100-1:500
Type: Primary
Antigen: PRKAB1 (protein kinase, AMP-activated, beta 1 non-catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: