Anti-HES5 Rabbit Polyclonal Antibody
BIRBORB95465-50
: Biorbyt
- Antibody type:Primary
- Antigen name:Hes family bHLH transcription factor 5
- Antigen symbol:HES5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage buffer:Tris-glycine, pH 7,4, 0,02% sodium azide, 0,5% BSA
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:50 µg
- Pk:50 µG
ELISA: 1:10000; Immunochemistry: 2.5 ?g/ml; Western Blot: 1:500-1:1000
Type: Primary
Antigen: HES5 (hes family bHLH transcription factor 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat