Anti-UCHL1 Rabbit Polyclonal Antibody
Catalog # BIRBORB95370-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin Carboxyl-Terminal Esterase L1
- Antigen symbol:UCHL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Storage buffer:PBS, 0,2% gelatin, 0,05% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunochemistry: 10 ?g/ml; Western Blot: 0.1-1 ?g/ml
Type: Primary
Antigen: UCHL1 (ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: