Anti-A2M Rabbit Polyclonal Antibody
Catalog # BIRBORB88687-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Alpha-2-macroglobulin
- Antigen symbol:A2M
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Western blot:Yes
- Antigen synonyms:CPAMD5
- Storage buffer:PBS, 0,01% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunochemistry: 10 ?g/ml; Western Blot: 1 ?g/ml
Type: Primary
Antigen: A2M (alpha-2-macroglobulin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: