Anti-LDB2 Rabbit Polyclonal Antibody
Catalog # BIRBORB87405-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:LIM domain binding 2
- Antigen symbol:LDB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Western blot:Yes
- Storage buffer:0,02 M potassium phosphate, 0,15 M sodium chloride, pH 7,2, 0,01% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
ELISA: 1:5000-1:20000; Immunochemistry: 5 ?g/ml; Western Blot: 1:500-1:2000
Type: Primary
Antigen: LDB2 (LIM domain binding 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: