Anti-CA2 Rabbit Polyclonal Antibody
Catalog # BIRBORB87394-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:carbonic anhydrase II
- Antigen symbol:CA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Storage buffer:PBS, pH 7,2, 0,01% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: CA2 (carbonic anhydrase II)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: