Anti-HTT Rabbit Polyclonal Antibody
Catalog # BIRBORB86459-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:huntingtin
- Antigen symbol:HTT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage buffer:0,02 M potassium phosphate, 0,15 M sodium chloride, pH 7,2, 0,01% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide from human HTT/Huntingtin
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
ELISA: 1:10000-1:40000; Immunochemistry: 10-20 ?g/ml; Western Blot: 1:500-1:3000
Type: Primary
Antigen: HTT (huntingtin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat