Anti-LRRC15 Rabbit Polyclonal Antibody
100 µg
BIRBORB78434-100
Marketsource
: Biorbyt
ELISA: 1:20000-1:80000; Western Blot: 1:1000-1:2000
Type: Primary
Antigen: LRRC15 (leucine rich repeat containing 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Size:100 µg
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by affinity chromatography
- Antibody type:Primary
- Isotype:IgG
- Storage temperature:Cool
- Host:Rabbit
- Shipping temperature:Ice
- Antigen name:leucine rich repeat containing 15
- Cat. No.:BIRBORB78434-100
- Reactivity:Human
- Immunogen:Recombinant protein
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:LRRC15
- ELISA:Yes



