Anti-ADAMTS2 Rabbit Polyclonal Antibody
Catalog # BIRBORB76079-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:ADAM metallopeptidase with thrombospondin type 1 motif, 2
- Antigen symbol:ADAMTS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Form:lyophilized
- Amino acid number:1189 to 1204
- Storage buffer:5mg BSA, 0,9mg NaCl, 0,2mg Na2HPO4, 0,05mg Thimerosal, 0,05mg NaN3.
- Sequence:KTRNQRIQELIDEMRK
- Storage temperature:Cool
- Concentration:500ug/ml,
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human ADAMTS2(1189-1204aa KTRNQRIQELIDEMRK), different from the related rat and mouse sequences by three amino acids.
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
0.1-0.5 ?g/ml
Type: Primary
Antigen: ADAMTS2 (ADAM metallopeptidase with thrombospondin type 1 motif, 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: