Anti-PRICKLE1 Rabbit Polyclonal Antibody
BIRBORB75688-100
: Biorbyt
- Antibody type:Primary
- Antigen name:prickle homolog 1 (Drosophila)
- Antigen symbol:PRICKLE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage temperature:Cool
- Concentration:1 mg/mL
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: PRICKLE1 (prickle homolog 1 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat