Anti-BAIAP2L1 Rabbit Polyclonal Antibody
Catalog # BIRBORB69891-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:BAI1-associated Protein 2-like 1
- Antigen symbol:BAIAP2L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Concentration:0,55 mg/ml
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: BAIAP2L1 (BAI1-associated protein 2-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human