Order Entry
Italy
ContactUsLinkComponent
Anti-PRKAG2 Rabbit Polyclonal Antibody
Anti-PRKAG2 Rabbit Polyclonal Antibody
Catalog # BIRBORB69809-100
Supplier:  Biorbyt
undefined
Anti-PRKAG2 Rabbit Polyclonal Antibody
Catalog # BIRBORB69809-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein Kinase AMP-Activated Non-Catalytic Subunit Gamma 2
  • Antigen symbol:
    PRKAG2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Concentration:
    0,96mg/ml
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: PRKAG2 (protein kinase, AMP-activated, gamma 2 non-catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human