Anti-CA9 Mouse monoclonal antibody unconjugated [clone: 2D3]
Catalog # BIRBORB69359-100
Nome del fornitore: Biorbyt
Specifiche
- Antibody type:Primary
- Antigen name:carbonic anhydrase IX
- Antigen symbol:CA9
- Clonality:Monoclonal
- Clone:2D3
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Format:ascites fluid
- Storage buffer:0,03% sodium azide.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Purified recombinant fragment of human CA9 expressed in E. Coli.
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifiche
Informazioni su questo articolo
Western Blot: 1:500-1:2000; Immunochemistry: 1:200-1:1000; Flow Cytometry: 1:200-1:400; ELISA: 1:10000
Type: Primary
Antigen: CA9 (carbonic anhydrase IX)
Clonality: monoclonal
Clone: 2D3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human