Order Entry
Italy
ContactUsLinkComponent
Anti-MUM1 Mouse Monoclonal Antibody [clone: 4G10]
Anti-MUM1 Mouse Monoclonal Antibody [clone: 4G10]
Catalog # BIRBORB69264-100
Supplier:  Biorbyt
undefined
Anti-MUM1 Mouse Monoclonal Antibody [clone: 4G10]
Catalog # BIRBORB69264-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Melanoma Associated Antigen (Mutated) 1
  • Antigen symbol:
    MUM1
  • Clonality:
    Monoclonal
  • Clone:
    4G10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Format:
    ascites fluid
  • Storage buffer:
    0,03% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Purified recombinant fragment of human MUM1 expressed in E. Coli.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Western Blot: 1:500-1:2000; Immunochemistry: 1:200-1:1000; ELISA: 1:10000

Type: Primary
Antigen: MUM1 (melanoma associated antigen (mutated) 1)
Clonality: monoclonal
Clone: 4G10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human