Anti-LPA Mouse Monoclonal Antibody
Catalog # BIRBORB69132-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:lipoprotein, Lp(a)
- Antigen symbol:LPA
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Format:ascites fluid
- Storage buffer:0,03% sodium azide.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Purified recombinant fragment of LPA (4330-4521) expressed in E. Coli.
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Western Blot: 1:500-1:2000; Immunochemistry: 1:200-1:1000; ELISA: 1:10000
Type: Primary
Antigen: LPA (lipoprotein, Lp(a))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human