Anti-Methylated Lysine Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB67536-400
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Methylated Lysine
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Storage buffer:PBS, 50% glycerol, 0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Methylated KLH
- Purification:purified by affinity chromatography
- Size:400 µg
- Pk:400 µG
Specifications
About this item
Western Blot: 1:2000-1:5000
Type: Primary
Antigen: Methylated Lysine
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: