Anti-Mage-1/Hla-A1 Protein Rabbit Polyclonal Antibody (Biotin)
Catalog # BIRBORB114245-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Hla-A1 Protein
- Antigen symbol:MAGE-1
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:34 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Mage-1/Hla-A1 Protein
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human