Anti-IL6 alpha Receptor Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
BIRBORB113697-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Interleukin 6 receptor alpha
- Antigen symbol:IL6RA
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:50 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: IL6 alpha Receptor
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat