Anti-Actin, beta cytoplasmic 1 Rabbit Polyclonal Antibody
Catalog # BIRBORB109434-200
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Actin beta
- Antigen symbol:ACTB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: Actin, beta cytoplasmic 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human