Anti-DNase II alpha Rabbit Polyclonal Antibody
Catalog # BIRBORB108712-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:DNase II alpha
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to amino acids 347 to 360 of human DNase II precursor (2-4).
- Purification:purified by antigen immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: DNase II alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human