Order Entry
Italy
ContactUsLinkComponent
Anti-p21 WAF-1 Mouse Monoclonal Antibody
Anti-p21 WAF-1 Mouse Monoclonal Antibody
Catalog # BIRBORB108547-100
Supplier:  Biorbyt
Anti-p21 WAF-1 Mouse Monoclonal Antibody
Catalog # BIRBORB108547-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    P21
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Antigen synonyms:
    WAF1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    A recombinant full length p21protein recognizing a 21kDa protein
  • Purification:
    purified by Protein A/G Chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: p21 WAF-1
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human