Order Entry
Italy
ContactUsLinkComponent
Anti-p44/42 MAP kinase Rabbit Polyclonal Antibody
Anti-p44/42 MAP kinase Rabbit Polyclonal Antibody
Catalog # BIRBORB106103-100
Supplier:  Biorbyt
undefined
Anti-p44/42 MAP kinase Rabbit Polyclonal Antibody
Catalog # BIRBORB106103-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    p44/42 MAP Kinase
  • Antigen symbol:
    p44/42 MAPK
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Modification:
    phospho-Thr202
  • Storage buffer:
    Phosphate buffer saline, pH. 7.4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Molecular weight:
    44+42 kDa
  • Storage temperature:
    Cool
  • Concentration:
    20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ul
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthesized peptide derived from human p44/42 MAP Kinase around the phosphorylation site of Threonine 202
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Western Blot: 1:500-1:2000

Type: Primary
Antigen: p44/42 MAP kinase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat