Anti-CA8 Rabbit polyclonal antibody
Nome del fornitore: Biorbyt
Certificati
Informazioni su questo articolo
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: CA8 (carbonic anhydrase VIII)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifiche
- Catalog No:
- BIRBORB101311-200
- BIRBORB101311-100
- Antigen symbol:
- CA8
- CA8
- Antigen name:
- carbonic anhydrase VIII
- carbonic anhydrase VIII
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- liquid
- liquid
- Molecular weight:
- 33 kDa
- 33 kDa
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Concentration:
- 0,5 mg/ml
- 0,5 mg/ml
- Storage buffer:
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifiche
Frequently Bought Together