Anti-LAT2 Rabbit Polyclonal Antibody
Catalog # BIRBORB44790-0.1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:linker for activation of T cells family, member 2
- Antigen symbol:LAT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: LAT2 (linker for activation of T cells family, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human