Order Entry
Italy
ContactUsLinkComponent
Anti-SIT1 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Anti-SIT1 Mouse Monoclonal Antibody (PE (Phycoerythrin))
  BIRBORB44489-0.1
 :  Biorbyt
Anti-SIT1 Mouse Monoclonal Antibody (PE (Phycoerythrin))
  BIRBORB44489-0.1
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen name:
    Signaling threshold regulating transmembrane adaptor 1
  • Antigen symbol:
    SIT1
  • Clonality:
    Monoclonal
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μg
  • Pk:
    0,1 mg

 

 

Type: Primary
Antigen: SIT1 (signaling threshold regulating transmembrane adaptor 1)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human